missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper Thinfoil, Oxygen-Free High Conductivity (OFHC), 0.008mm (0.0003in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 045943FI
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper Thinfoil | |
7440-50-8 | |
Cu | |
50 x 50 mm | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
99.99% | |
≈0.04g/25x25mm |
Oxygen-Free High Conductivity (OFHC), 0.008mm (0.0003 in.) thick | |
(metals basis) | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
Odorless | |
Thin Foil |
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Safety and Handling
EINECSNumber : 231-159-6
RTECSNumber : GL5325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only