missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Thinfoil, 0.0125mm (0.0005in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.07g/25 x 25mm; Not certified pinhole free

Supplier:  Thermo Scientific Chemicals 038381FI

 View more versions of this product

Catalog No. AA38381FI



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper
Copper colored thinfoil
Cu
50 x 50 mm
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Odorless
7440-50-8
99.9% (Metals basis)
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
63.55
CHEBI:30052

Chemical Identifiers

7440-50-8
63.55
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only