Please login to your online account to display your discounted pricing

Copper Sulfate - Sulfamic Acid, For Dissolved Oxygen, LabChem™

$101.91 - $101.91

Chemical Identifiers

Chemical Name or Material Copper Sulfate - Sulfamic Acid
CAS 7732-18-5
CAS Max % 89.98
CAS 7758-99-8
CAS Max % 4.63
CAS 5329-14-6
CAS Max % 2.96
CAS 64-19-7
CAS Max % 2.43
Name Note For Dissolved Oxygen
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC134702

 
lab chem
LC134702
Poly Bottle 1L Each for $101.91
Description & Specifications

Specifications

Formula Weight 249.68
Chemical Name or Material Copper Sulfate - Sulfamic Acid
CAS Max % 2.96
CAS 64-19-7
CAS Max % 2.43
Packaging Poly Bottle
Specific Gravity 1.08g/mL
Name Note For Dissolved Oxygen
Physical Form Liquid
Density 1.08g/mL
Linear Formula CuSO4·5H2O
Color Clear, blue
CAS 7732-18-5
UN Number UN1760
Quantity 1L
Solubility Information Soluble in water
Health Hazard 1 Danger
CAS Max % 89.98
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
CAS 7758-99-8
Health Hazard 3 GHS P Statement
Do not breathe mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
CAS Max % 4.63
Recommended Storage Room Temperature
CAS 5329-14-6
Safety and Handling

Danger

GHS P Statement
Do not breathe mist.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

Room Temperature