Please login to your online account to display your discounted pricing

Copper Sulfate Solution (For AOAC Method 930.28), Fisher Chemical


Quantity 1L
Packaging Poly Bottle

Manufacturer: ricca chemical company  SC92021

 View more versions of this product

Catalog No. SC9202-1

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1L
Methods For AOAC Method 930.28
Chemical Name or Material Copper Sulfate Solution
CAS 7732-18-5
CAS Min % 92.0
CAS Max % 95.0
Packaging Poly Bottle
ChemAlert Storage Symbol Gray
CAS 7758-99-8
CAS Min % 6.35
CAS Max % 6.61

Specifications

Chemical Name or Material Copper Sulfate Solution
CAS 7732-18-5
CAS Min % 92.0
CAS Max % 95.0
CAS 7758-99-8
CAS Min % 6.35
CAS Max % 6.61
Accessories