Please login to your online account to display your discounted pricing

Copper Sulfate, Pentahydrate, ACS, LabChem™

$94.83 - $94.83

Chemical Identifiers

Chemical Name or Material Copper Sulfate
CAS 7758-99-8
CAS Max % 100
Name Note Pentahydrate
Purity Grade ACS Grade
Linear Formula CuSO4·5H2O
Formula Weight 249.68
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC134051

 
lab chem
LC134051
500g Each for $94.83
Description & Specifications

Specifications

Assay 98.0 to 102.0%
Formula Weight 249.68
Chemical Name or Material Copper Sulfate
EINECS Number 231-847-6
Identification Passes Test
Name Note Pentahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Solid
Solubility Soluble in water
Specific Gravity 2286kg/m3
Storage Information Room Temperature
Linear Formula CuSO4·5H2O
CAS 7758-99-8
UN Number UN3077
Decomposition Information Fume; Sulfur compounds
Color fine blue crystals
Quantity 500g
CAS Max % 100
Density 2286kg/m3