Please login to your online account to display your discounted pricing

Copper Sulfate - Hydrochloric Acid, Certified, LabChem™

$130.71 - $130.71

Chemical Identifiers

Chemical Name or Material Copper Sulfate - Hydrochloric Acid
CAS 7732-18-5
CAS Max % 77.4
CAS 7758-99-8
CAS Max % 17.8
CAS 7647-01-0
CAS Max % 4.8
Purity Grade Certified Grade
Linear Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC134504

 
lab chem
LC134504
4L Each for $130.71
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 249.68
Chemical Name or Material Copper Sulfate - Hydrochloric Acid
CAS Max % 4.8
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1.1g/mL
Storage Information Room Temperature
Linear Formula CuSO4·5H2O
CAS 7732-18-5
UN Number UN1789
Decomposition Information Sulfur compounds; Copper; Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Color Clear, blue
Quantity 4L
Health Hazard 1 Danger
CAS Max % 77.4
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
CAS 7758-99-8
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Store locked up.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 17.8
Density 1.1g/mL
CAS 7647-01-0
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Store locked up.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.