missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper Sulfate, For Nitrogen (Nitrate), Certified, 2.0% (w/v) ±0.1%, LabChem™
Supplier: LabChem LC134201
Specifications
Clear, blue | |
Poly Bottle | |
Room Temperature |
Liquid | |
500 mL | |
Copper Sulfate |