Please login to your online account to display your discounted pricing

Copper Sulfate, For Morpholine, 2g/L, LabChem™

$54.37 - $54.37

Chemical Identifiers

Chemical Name or Material Copper Sulfate
CAS 7732-18-5
CAS Max % 99.8
CAS 7758-99-8
CAS Max % 0.2
Name Note For Morpholine
Linear Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC134101

 
lab chem
LC134101
500mL Each for $54.37
Description & Specifications

Specifications

Formula Weight 249.68
Chemical Name or Material Copper Sulfate
Decomposition Information Sulfur compounds
Color Clear, blue
Quantity 500mL
Name Note For Morpholine
Physical Form Liquid
Concentration 2g/L
Linear Formula CuSO4·5H2O
Density 1g/mL
CAS 7732-18-5
CAS Max % 99.8
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 7758-99-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.2
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.