missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Sulfate, Certified, 1.186 ±0.003, LabChem™

Manufacturer:  LabChem LC134454

 View more versions of this product

Catalog No. LC134454



Specifications

Specifications

Copper Sulfate
70.48,29.52
CuSO4·5H2O
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper;sulfate;pentahydrate
24463
249.68
Certified
Passes Test
Soluble in water
Room Temperature
Blue
7758-99-8,7732-18-5
CuH10O9S
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
249.677
CHEBI:31440
Liquid
1.186 ±0.003
Poly Bottle
1.186 at 18.4°C
Sulfur compounds; Copper
4L

Chemical Identifiers

7758-99-8
249.677
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
CHEBI:31440
O.O.O.O.O.[O-]S(=O)(=O)[O-].[Cu+2]
CuH10O9S
JZCCFEFSEZPSOG-UHFFFAOYSA-L
24463
copper;sulfate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Do not eat, drink or smoke when using this product.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Collect spillage.

Warning

Documents
Promotions

Promotions