Please login to your online account to display your discounted pricing

Copper Sulfate, Certified, 0.0200M ±0.0002M (0.02N), LabChem™

$40.97 - $40.97

Chemical Identifiers

Chemical Name or Material Copper Sulfate
CAS 7732-18-5
CAS Max % 99.5
CAS 7758-99-8
CAS Max % 0.5
Purity Grade Certified Grade
Linear Formula CuSO4·5H2O
Formula Weight 249.68
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC134432

 
lab chem
LC134432
1L Each for $40.97
Description & Specifications

Specifications

Formula Weight 249.68
Purity Grade Certified Grade
Chemical Name or Material Copper Sulfate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Linear Formula CuSO4·5H2O
CAS 7732-18-5
Decomposition Information Sulfur compounds
Color Clear, blue
Quantity 1L
Traceability to NIST Traceable to NIST
CAS Max % 99.5
Health Hazard 2 GHS H Statement
Harmful to aquatic life with long lasting effects.
CAS 7758-99-8
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.5
Concentration 0.0200M ±0.0002M (0.02N)
Density 1g/mL
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life with long lasting effects.