missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Sulfate, Certified, 0.0100M ±0.0001M (0.01N), LabChem™

$46.31 - $46.31

Chemical Identifiers

CAS 7732-18-5,7758-99-8
CAS Max % 99.75,0.25
Linear Formula CuSO4·5H2O
Formula Weight 249.68
Chemical Name or Material Copper Sulfate
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC134422 Lab Chem
LC134422
1L Each for $46.31
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7758-99-8
CuSO4·5H2O
Copper Sulfate
99.75,0.25
249.68
Certified Grade

Specifications

249.68
7758-99-8,7732-18-5
Soluble in water
1g/mL
Poly Bottle
CuSO4·5H2O
Clear, blue
Sulfur compounds
Copper Sulfate
Liquid
Passes Test
1L
Traceable to NIST
99.75,0.25
Room Temperature
1g/mL
0.0100M ±0.0001M (0.01N)
Certified Grade

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.