Please login to your online account to display your discounted pricing

Copper Sulfate, 1% (w/v) Aqueous Solution, Ricca Chemical

$59.09 - $59.09

Chemical Identifiers

Chemical Name or Material Copper Sulfate
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7758-99-8
CAS Min % 0.97
CAS Max % 1.01
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R23168001A

 
ricca chemical company
RIR23168001A
Poly natural 1L Each for $59.09

R2316800500

 
ricca chemical company
RIR2316800500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear blue
Quantity 1L
CAS Max % 100.0
CAS Min % 97.0
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous Solution
Chemical Name or Material Copper Sulfate
Shelf Life 24 months
CAS 7732-18-5
CAS 7758-99-8
CAS Min % 0.97
Packaging Poly natural
CAS Max % 1.01
Safety and Handling

24 months