Please login to your online account to display your discounted pricing

Copper standard, Ricca Chemical

$68.68 - $68.68

Chemical Identifiers

Chemical Name or Material Copper standard
CAS 7732-18-5
CAS 7758-99-8
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

22-951-6

 
ricca chemical company
RI229516
Poly natural 500mL Each for $68.68

RR22853004A

 
ricca chemical company
RIR22853004A
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
CAS 7732-18-5
CAS 7758-99-8
Packaging Poly natural
Chemical Name or Material Copper standard