missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper Standard For ICP, 10,000μg/mL in 5% HNO3, Assurance Grade SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$407.18 - $801.29
Chemical Identifiers
CAS | 7440-50-8 |
---|---|
Molecular Formula | Cu |
Molecular Weight (g/mol) | 63.55 |
MDL Number | MFCD00010965 |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
PubChem CID | 23978 |
ChEBI | CHEBI:30052 |
IUPAC Name | copper |
SMILES | [Cu] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cu | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
CHEBI:30052 | |
[Cu] |
Cu | |
MFCD00010965 | |
23978 | |
copper |
Specifications
7440-50-8,7697-37-2,7732-18-5 | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Cu | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
Assurance | |
Copper Standard |
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010965 | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
5% HNO3 |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only