Please login to your online account to display your discounted pricing

Copper Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

$101.00 - $101.00

Chemical Identifiers

Chemical Name or Material Copper Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
CAS 007440-50-8
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ICP-029

 
ultra scientific
ICP029
125mL Each for $101.00
Description & Specifications

Specifications

Quantity 125mL
Solvent or Matrix Water with dilute nitric acid
Analyte or Component Names Water,Nitric acid,Copper
CAS Max % 2.0
CAS 007440-50-8
Chemical Name or Material Copper Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 0.1
Concentration or Composition (by Analyte or Components) 1000μg/mL