Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis)

$898.03 - $898.03

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40841KS

  SDS
alfa aesar™
40841KS
1 each Each for $898.03
Description & Specifications

Specifications

Chemical Name or Material Copper sputtering target
Quantity 1 each
Assay 99.999%
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay Percent Notes (metals basis)
CAS 7440-50-8
Odor Odorless
MDL Number MFCD00010965