Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.995% (metals basis)

$560.51 - $560.51

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43041KS

  SDS
alfa aesar™
43041KS
1 each Each for $560.51
Description & Specifications

Specifications

Assay 99.995%
CAS 7440-50-8
Odor Odorless
Chemical Name or Material Copper sputtering target
Quantity 1 each
Assay Percent Notes (metals basis)
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010965