missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)

$625.30 - $625.30

Chemical Identifiers

CAS 7440-50-8
MDL Number MFCD00010965
Chemical Name or Material Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40840KS SDS Alfa Aesar™
40840KS
1 each Each for $625.30
Specifications Description & Specifications

Chemical Identifiers

7440-50-8
MFCD00010965
Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick

Specifications

1 each
MFCD00010965
Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
7440-50-8
Odorless
99.999%, (metals basis)

Safety and Handling

TSCA : TSCA