Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis)

$569.55 - $569.55

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43040KS

  SDS
alfa aesar™
43040KS
1 each Each for $569.55
Description & Specifications

Specifications

Assay 99.995%
Odor Odorless
MDL Number MFCD00010965
CAS 7440-50-8
TSCA TSCA
Assay Percent Notes (metals basis)
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Chemical Name or Material Copper sputtering target
Quantity 1 each