Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis)

$659.96 - $659.96

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40839KS

  SDS
alfa aesar™
40839KS
1 each Each for $659.96
Description & Specifications

Specifications

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Odor Odorless
Assay 99.999%
MDL Number MFCD00010965
Quantity 1 each
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay Percent Notes (metals basis)
TSCA TSCA