Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.995% (metals basis)

$452.03 - $452.03

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43039KS

  SDS
alfa aesar™
43039KS
1 each Each for $452.03
Description & Specifications

Specifications

Chemical Name or Material Copper sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
Odor Odorless
Quantity 1 each