Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)

Magnetron sputtering source

$238.59 - $238.59

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40838KS

  SDS
alfa aesar™
40838KS
1 each Each for $238.59
Description & Specifications

Specifications

Chemical Name or Material Copper sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
Odor Odorless
RTECS Number GL5325000
Quantity 1 each