Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis)

Magnetron sputtering source

$364.64 - $364.64

Chemical Identifiers

Chemical Name or Material Copper sputtering target
CAS 7440-50-8
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43038KS

  SDS
alfa aesar™
43038KS
1 each Each for $364.64
Description & Specifications

Specifications

Quantity 1 each
TSCA TSCA
CAS 7440-50-8
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
Odor Odorless
Assay 99.995%
Chemical Name or Material Copper sputtering target