missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, 99.995+% (metals basis), Oxygen free, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04425818
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper slug | |
7440-50-8 | |
Cu | |
50 g | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
≥99.995% | |
≈0.45g/slug |
3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Oxygen free | |
(metals basis) | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
Odorless | |
Slug |
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Safety and Handling
EINECSNumber : 231-159-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only