Please login to your online account to display your discounted pricing

Copper silicide, Silver-colored pieces, 99.5% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Copper silicide
CAS 12159-07-8
Physical Form Pieces
Assay Percent Range 99.5% (Metals basis)
Molecular Formula Cu5Si
Formula Weight 345.79
MDL Number MFCD00134719
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1284414

 
alfa aesar
1284414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1284422

 
alfa aesar
1284422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Pieces
Formula Weight 345.79
Quantity 25g
Color Silver
Chemical Name or Material Copper silicide
EINECS Number 235-286-8
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
Molecular Formula Cu5Si
MDL Number MFCD00134719
CAS 12159-07-8
Health Hazard 1 H315-H319-H335; Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335; Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.