missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper shot, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4421336

 View more versions of this product

Catalog No. AA4421336



Specifications

Specifications

Copper
7440-50-8
8.94g/cm3
Cu
anode
[Cu]
Odorless
23978
Shot, 0.8-2mm (0.03-0.08 in.)
Undesignated
500g
Copper colored shot
99.5% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
0 hPa

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents