missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Shot, 6mm (0.24 in.) & down, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1095514

 View more versions of this product

Catalog No. AA1095514



Specifications

Specifications

Copper
7440-50-8
99.999% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
Undesignated
25g
Copper colored shot
Cu
anode
[Cu]
Odorless
23978
Shot, 6mm (0.24 in.) and down

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Documents