missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Shot, 0.6-0.8mm (0.02-0.03 in.), 99.5% (Metals basis), Alfa Aesar™

$98.66 - $98.66

Chemical Identifiers

CAS 7440-50-8
Physical Form Shot, 0.6-0.8mm (0.02-0.03 in.)
Assay Percent Range 99.5% (Metals basis)
MDL Number MFCD00010965
Chemical Name or Material Copper
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44212A3 SDS Alfa Aesar
44212A3
2kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA4421236 SDS Alfa Aesar
4421236*
500g Each for $98.66
Specifications Description & Specifications

Chemical Identifiers

7440-50-8
99.5% (Metals basis)
Copper
Shot, 0.6-0.8mm (0.02-0.03 in.)
MFCD00010965

Specifications

2kg
Shot, 0.6-0.8mm (0.02-0.03 in.)
Copper colored shot
7440-50-8
Copper
Copper colored
99.5% (Metals basis)
Odorless
MFCD00010965