Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper rod, Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia. 183cm

Chemical Identifiers

Chemical Name or Material Copper rod
Name Note Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia.
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45189AEX

  SDS
alfa aesar™
45189AEX
183cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Name Note Oxygen-Free High Conductivity (OFHC), alloy 101, 9.53mm (0.375 in.) dia.
MDL Number MFCD00010965
Chemical Name or Material Copper rod
Quantity 183cm
Odor Odorless