Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper rod, Oxygen-Free High Conductivity (OFHC), alloy 101, 3.175mm (0.125 in.) dia. 183cm

Chemical Identifiers

Chemical Name or Material Copper rod
Name Note Oxygen-Free High Conductivity (OFHC), alloy 101, 3.175mm (0.125 in.) dia.
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45146ABR

  SDS
alfa aesar™
45146ABR
183cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper rod
TSCA TSCA
Name Note Oxygen-Free High Conductivity (OFHC), alloy 101, 3.175mm (0.125 in.) dia.
MDL Number MFCD00010965
Odor Odorless
Quantity 183cm