missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper rod, 9.5mm (0.4 in.) dia., Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1015636

 View more versions of this product

Catalog No. AA1015636



Specifications

Specifications

Copper rod
500g
7440-50-8
Cu
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
[Cu]
Odorless
23978
Rod, ≈6.33g/cm
Puratronic™
≈6.33g/cm
9.5mm (0.4 in.) dia.
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.999%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents