missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Copper rod, 19mm (0.8 in.) dia., Puratronic™, 99.999% (metals basis)
Manufacturer: Alfa Aesar™ 12754DJ
Specifications
Copper rod | |
50mm | |
7440-50-8 | |
Cu | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
Odorless | |
23978 | |
Rod | |
Puratronic™ |
≈25.3g/cm | |
19mm (0.8 in.) dia. | |
(metals basis) | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
63.546 | |
CHEBI:30052 | |
99.999% |
Chemical Identifiers
7440-50-8 | |
63.546 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Safety and Handling
TSCA : TSCA