Please login to your online account to display your discounted pricing

Copper, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Cu) Solute: Copper Nitrate Solvent: Dilute Nitric Acid

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SC194500

 View more versions of this product

Catalog No. SC194-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Copper, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 19004-19-4
ChemAlert Storage Symbol White
CAS 7697-37-2
CAS 7732-18-5
DOT Information DOT Class 8, : Corrosive
Purity Grade Certified Grade
Physical Form Liquid
Concentration 990 to 1010ppm
MDL Number MFCD00010965
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
Traceability to NIST 3114 COPPER REF. STD.
Vapor Pressure 14mmHg at 20°C
Boiling Point 100°C
Color Blue
Quantity 500mL

Specifications

Chemical Name or Material Copper, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 19004-19-4
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010965
Accessories
 

SDS & Safety Documents