missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper powder, spherical, -170+270 mesh, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 45544A3

 View more versions of this product

Catalog No. AA45544A3



Specifications

Specifications

Copper powder
7440-50-8
Cu
UN3089
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
Odorless
copper
23978
Spherical
Red
2kg
(metals basis)
MFCD00010965
−170+270 Mesh
RYGMFSIKBFXOCR-UHFFFAOYSA-N
[Cu]
63.546
CHEBI:30052
99.9%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : GL5325000

TSCA : TSCA

SDS Documents