Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper powder, spherical, -100 mesh, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Copper powder
CAS 7440-50-8
MDL Number MFCD00010965
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1107030

  SDS
alfa aesar™
1107030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11070A1

  SDS
alfa aesar™
11070A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11070D9

  SDS
alfa aesar™
11070D9
5 x 1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Sperical Powder
Color Copper
MDL Number MFCD00010965
Assay Percent Notes (metals basis)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Odor Odorless
TSCA TSCA
Mesh Size −100 Mesh
Quantity 250g
RTECS Number GL5325000
UN Number UN3077
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 7440-50-8
Chemical Name or Material Copper powder
Assay 99.5%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.