Learn More
Copper powder, semispherical, -50+70 mesh, 99% (metals basis), Thermo Scientific Chemicals
$81.53 - $81.53
Chemical Identifiers
CAS | 7440-50-8 |
---|---|
Molecular Formula | Cu |
Molecular Weight (g/mol) | 63.55 |
MDL Number | MFCD00010965 |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
Synonym | cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer |
PubChem CID | 23978 |
ChEBI | CHEBI:30052 |
IUPAC Name | copper |
SMILES | [Cu] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4503022
|
Thermo Scientific Chemicals
04503022 |
100 g |
Each for $81.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-50-8 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Specifications
7440-50-8 | |
MFCD00010965 | |
100 g | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
23978 | |
99% | |
Odorless | |
(metals basis) |
Cu | |
UN3077 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
63.55 | |
CHEBI:30052 | |
−50+70 Mesh | |
Copper powder |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319-H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 231-159-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only