Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper powder, -22 mesh, Puratronic™, 99.999% (metals basis)

$197.03 - $197.03

Chemical Identifiers

Chemical Name or Material Copper powder
CAS 7440-50-8
Purity Grade Puratronic™
MDL Number MFCD00010965
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1060909

  SDS
alfa aesar™
1060909
10g Each for $197.03

AA1060918

  SDS
alfa aesar™
1060918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper powder
TSCA TSCA
Purity Grade Puratronic™
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
UN Number UN3077
Mesh Size −22 Mesh
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Copper
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number GL5325000
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.