missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Powder, -150 mesh, 99.5% (Metal Basis), Alfa Aesar™

Apparent density 2.6-2.8g/cm3

Manufacturer:  Thermo Fisher Scientific 10160A3

 View more versions of this product

Catalog No. AA10160A3



Specifications

Specifications

Copper
7440-50-8
Cu
-150 Mesh
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Powder
2kg
99.5%, metal basis
MFCD00010965
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
[Cu]
63.546
CHEBI:30052

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode, blister, bronze, cathode, cobre, cuivre, cuprum, kupfer, powder, precipitates
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335-H500

DOTInformation : DOT Class: 4.1; Packaging Group: III

EINECSNumber : 231-159-6

RTECSNumber : GL5325000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents