missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in.) thick, , Alfa Aesar™

Manufacturer:  Alfa Aesar 45190GT

 View more versions of this product

Catalog No. AA45190GT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Copper
7697-37-2
HNO3
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
Odorless
944
Plate, Oxygen-Free High Conductivity (Ofhc), Alloy 101, 6.35mm (0.25 in.) thick
150 x 150mm
Copper colored plate
MFCD00010965
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107
Undesignated

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00010965
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]
SDS Documents