Please login to your online account to display your discounted pricing

Copper Plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 6.35mm (0.25in.) thick, , Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Copper
Physical Form Plate, Oxygen-Free High Conductivity (Ofhc), Alloy 101, 6.35mm (0.25 in.) thick
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45190HB

  SDS
alfa aesar
45190HB
300 x 300mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA45190GT

  SDS
alfa aesar
45190GT
150 x 150mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Plate, Oxygen-Free High Conductivity (Ofhc), Alloy 101, 6.35mm (0.25 in.) thick
Quantity 300 x 300mm
Color Copper colored
Chemical Name or Material Copper
MDL Number MFCD00010965
Odor Odorless
Appearance Copper colored plate