missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Plate, 6.35mm (0.25 in.) thick, 99.9% (Metals basis), Alfa Aesar™

Form: Approx. 569g/10 x 10cm

Manufacturer:  Alfa Aesar 13376CU

 View more versions of this product

Catalog No. AA13376CU



Specifications

Specifications

Copper
7440-50-8
99.9% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
Undesignated
20 x 20cm
Copper colored plate
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
Plate, 6.35mm (0.25 in.) thick

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Documents