missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Copper Plate, 6.35mm (0.25 in.) thick, 99.9% (Metals basis), Alfa Aesar™
Form: Approx. 569g/10 x 10cm
Manufacturer: Alfa Aesar 13376RR
Specifications
Copper | |
7440-50-8 | |
8.94g/cm3 | |
Cu | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
Odorless | |
23978 | |
Plate, 6.35mm (0.25 in.) thick |
10 x 10cm | |
Copper colored plate | |
99.9% (Metals basis) | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
63.546 | |
CHEBI:30052 | |
Undesignated |
Chemical Identifiers
7440-50-8 | |
63.546 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |