missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13867AD

 View more versions of this product

Catalog No. AA13867AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Copper
Clear liquid
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
Liquid
Colorless
7440-50-8,7440-50-8,7732-18-5
Cu
Nitric acid: 5.0%; Copper: 0.1%; Water: 94.9%
[Cu]
63.546
CHEBI:30052
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
CHEBI:30052
[Cu]
Cu
MFCD00010965
23978
copper
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents