missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper phthalocyanine, dye content ca 95%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 269150250

 View more versions of this product

Catalog No. AC269150250



Description

Description

Specifications

Specifications

Copper phthalocyanine
147-14-8
MFCD00010719
C.I. 74160, Pigment blue 15
576.08
≥94% (ex Cu)
Solubility in water: practically insoluble. Other solubilities: soluble in 98% h2so4, practically insoluble in alcohol and hydrocarbons
Blue
dye content ca 95%
C32H16CuN8
15, 2505
Authentic
Fine Crystalline Powder
Glass bottle
Complies
25g

Chemical Identifiers

147-14-8
C32H16CuN8
MFCD00010719
C.I. 74160, Pigment blue 15
Safety and Handling

Safety and Handling

EINECSNumber : 205-685-1

RTECSNumber : GL8510000

TSCA : TSCA

SDS Documents