missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Oxide Wire, ACS Grade, 0.65 x 6mm, For Elementary Analysis, MilliporeSigma™

Manufacturer:  MilliporeSigma 1027671000

 View more versions of this product

Catalog No. M1027671000



Description

Description

For elementary analysis
Specifications

Specifications

Copper oxide
1317-38-0
UN 3077
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
O=[Cu]
79.545
79.55
ACS
Glass bottle
Surface: CuO, Core: Cu2O
CuO
Dark gray wire (0.65 x 6mm)
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
Wire
Odorless
1kg

Chemical Identifiers

1317-38-0
79.545
cupric oxide, copper ii oxide, copper oxide, copper monoxide, copper oxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
oxocopper
CuO
QPLDLSVMHZLSFG-UHFFFAOYSA-N
14829
O=[Cu]
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.
H410: Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-269-1

Documents
Promotions

Promotions