Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper, Oil based standard solution, Specpure™, Cu 1000μg/g

Chemical Identifiers

Chemical Name or Material Copper
Name Note Oil based standard solution
Purity Grade Specpure™
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4727618

 
alfa aesar™
4727618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper
Quantity 50g
Name Note Oil based standard solution
Purity Grade Specpure™
Physical Form Liquid
Concentration Cu 1000µg/g
MDL Number MFCD00010965