Alfa Aesar™ Copper Nickel wire, 1.6mm (0.064 in.) dia.
Manufacturer:  Alfa Aesar™ 43273G5
 View more versions of this product
Catalog No. AA43273G5
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Cu:Ni 55:45 wt%
≈17.9g/m
1.6mm (0.064 in.) dia.
Wire
Copper Nickel wire
5m
MFCD00801098
Documents