missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper(II) Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T129225G

 View more versions of this product

Catalog No. T129225G



Specifications

Specifications

Copper(II) Trifluoromethanesulfonate
25g
CHCuF3O3S
1759
GZWXEFRPSWBAGC-UHFFFAOYSA-N
copper;trifluoromethanesulfonic acid
45051791
Crystal-Powder at 20°C
Undesignated
34946-82-2
MFCD00077492
copper; trifluoromethanesulfonic acid
C(F)(F)(F)S(=O)(=O)O.[Cu]
213.617
361.67
≥98.0% (T)

Chemical Identifiers

34946-82-2
213.617
GZWXEFRPSWBAGC-UHFFFAOYSA-N
45051791
C(F)(F)(F)S(=O)(=O)O.[Cu]
CHCuF3O3S
MFCD00077492
copper; trifluoromethanesulfonic acid
copper;trifluoromethanesulfonic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents