missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) trifluoromethanesulfonate, 99%

CAS: 34946-82-2 | CHCuF3O3S | 213.617 g/mol

Supplier:  Thermo Scientific Chemicals 04013318

 View more versions of this product

Catalog No. AA4013318


Only null left
Explore available promotions

Description

Description

Copper(II) trifluoromethanesulfonate is a mild lewis acid. It is used as catalyst which promotes dehydration of alcohols and diols to alkenes at ambient temperatures. It is widely used to generate carbenoid species from ?-diazo esters and ketones, via in situ reduction to the Cu(I) species. It is also promotes the reaction between diazo esters and imines to give aziridines. It catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes, Friedel-Crafts alkylation, acylation reactions of aromatics and addition of trimethylsilyl cyanide to carbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Copper(II) trifluoromethanesulfonate
34946-82-2
Powder
CHCuF3O3S
UN3261
copper; trifluoromethanesulfonic acid
GZWXEFRPSWBAGC-UHFFFAOYSA-N
copper;trifluoromethanesulfonic acid
45051791
≤99%
(decomposition)
Blue to White
50 g
MFCD00077492
4028198
Soluble in water.
C(F)(F)(F)S(=O)(=O)O.[Cu]
213.617
361.68
Hygroscopic

Chemical Identifiers

34946-82-2
213.617
GZWXEFRPSWBAGC-UHFFFAOYSA-N
45051791
C(F)(F)(F)S(=O)(=O)O.[Cu]
CHCuF3O3S
MFCD00077492
copper; trifluoromethanesulfonic acid
copper;trifluoromethanesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 252-300-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only