missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper (II) tetrafluoroborate hexahydrate, 98%, Alfa Aesar

Manufacturer:  Alfa Aesar 2612736

 View more versions of this product

Catalog No. AA2612736



Specifications

Specifications

Copper(II) tetrafluoroborate hexahydrate
72259-10-0
B2CuF8H12O6
Blue crystals
FBYIXFBIARQMJJ-UHFFFAOYSA-N
copper;ditetrafluoroborate;hexahydrate
15774553
Crystalline
500g
hexahydrate
998%
MFCD00150223
copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci
[B-](F)(F)(F)F.[B-](F)(F)(F)F.O.O.O.O.O.O.[Cu+2]
345.243
345.25; 237.15 (anhydrous)
Blue

Chemical Identifiers

72259-10-0
345.243
FBYIXFBIARQMJJ-UHFFFAOYSA-N
15774553
[B-](F)(F)(F)F.[B-](F)(F)(F)F.O.O.O.O.O.O.[Cu+2]
B2CuF8H12O6
MFCD00150223
copper ii tetrafluoroborate hexahydrate, copper 2+ hexahydrate ditetrafluoroborate, copper ditetrafluoroborate hexahydrate, copper 2+ tetrafluoroborate-water 1/2/6, copper 2+ ion hexahydrate ditetrafluoroborate, borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci
copper;ditetrafluoroborate;hexahydrate
Safety and Handling

Safety and Handling

H314-H318; Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 253-959-4

storageNote1 : Hygroscopic

SDS Documents